Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGTGGAATTTTTTGCTGTCACAGA[A/G]TGTCCTGCAATAAATATCTCTGCAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607471 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BCAS4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BCAS4 - breast carcinoma amplified sequence 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001010974.2 | Intron | NP_001010974.1 | ||||
NM_017843.4 | Intron | NP_060313.3 | ||||
NM_198799.3 | Intron | NP_942094.2 | ||||
XM_011528886.2 | Intron | XP_011527188.1 | ||||
XM_011528887.2 | Intron | XP_011527189.1 | ||||
XM_017027932.1 | Intron | XP_016883421.1 |