Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATGAGTTTGTTAAGTGATTTTGAG[A/G]ATTTTGATGTGTAAATGCAGTTTTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
RTFDC1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RTFDC1 - replication termination factor 2 domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001283035.1 | Intron | NP_001269964.1 | ||||
NM_001283036.1 | Intron | NP_001269965.1 | ||||
NM_001283037.1 | Intron | NP_001269966.1 | ||||
NM_016407.4 | Intron | NP_057491.2 | ||||
XM_017027872.1 | Intron | XP_016883361.1 |