Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCAGTCAGAGGGGGAGATGTCATG[A/G]GTGCAATTCTCACATATGCCACAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605114 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SPO11 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SPO11 - SPO11, initiator of meiotic double stranded breaks | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012444.2 | Intron | NP_036576.1 | ||||
NM_198265.1 | Intron | NP_937998.1 | ||||
XM_005260379.3 | Intron | XP_005260436.1 | ||||
XM_005260380.3 | Intron | XP_005260437.1 | ||||
XM_005260381.3 | Intron | XP_005260438.1 | ||||
XM_005260382.4 | Intron | XP_005260439.1 | ||||
XM_011528756.2 | Intron | XP_011527058.1 | ||||
XM_011528757.2 | Intron | XP_011527059.1 |