Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTAAAAGTTCTAATTTTTTTGTTAC[G/T]TGCATTATCCTCTTTCCAGTGTTGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604896 MIM: 615958 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MKKS PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MKKS - McKusick-Kaufman syndrome | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLX4IP - SLX4 interacting protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001009608.2 | Intron | NP_001009608.1 | ||||
XM_017027647.1 | Intron | XP_016883136.1 | ||||
XM_017027648.1 | Intron | XP_016883137.1 | ||||
XM_017027649.1 | Intron | XP_016883138.1 | ||||
XM_017027650.1 | Intron | XP_016883139.1 | ||||
XM_017027651.1 | Intron | XP_016883140.1 | ||||
XM_017027652.1 | Intron | XP_016883141.1 | ||||
XM_017027653.1 | Intron | XP_016883142.1 |