Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAGAGATCCTGTTAGAGACTAGTT[C/T]TTTCCCCGCGCCACACACACACACG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612360 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ESF1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ESF1 - ESF1 nucleolar pre-rRNA processing protein homolog | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001276380.1 | 84 | Intron | NP_001263309.1 | |||
NM_016649.3 | 84 | Intron | NP_057733.2 | |||
XM_017027874.1 | 84 | UTR 5 | XP_016883363.1 |
NDUFAF5 - NADH:ubiquinone oxidoreductase complex assembly factor 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001039375.2 | 84 | Intron | NP_001034464.1 | |||
NM_024120.4 | 84 | Intron | NP_077025.2 | |||
XM_006723620.3 | 84 | Intron | XP_006723683.1 | |||
XM_006723622.2 | 84 | Intron | XP_006723685.1 | |||
XM_006723623.2 | 84 | Intron | XP_006723686.1 | |||
XM_006723624.2 | 84 | Intron | XP_006723687.1 | |||
XM_011529342.2 | 84 | Intron | XP_011527644.1 | |||
XM_011529344.2 | 84 | Intron | XP_011527646.1 | |||
XM_017028061.1 | 84 | Intron | XP_016883550.1 |