Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCAGGACCGTCTTCCTGCGGCAGC[A/G]CAGCCGCTTGCGAATCTGCTCCGTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607123 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PROKR2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PROKR2 - prokineticin receptor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_144773.3 | 1049 | Missense Mutation | CGC,TGC | R,C 268 | NP_658986.1 | |
XM_011529159.2 | 1049 | Missense Mutation | CGC,TGC | R,C 232 | XP_011527461.1 | |
XM_017027646.1 | 1049 | Missense Mutation | CGC,TGC | R,C 268 | XP_016883135.1 |