Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGTGAGACTGAGGGGCAAAGGAGA[A/G]AGGGCAGGGGCGTGTCTCAACCCTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613350 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC52A3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SLC52A3 - solute carrier family 52 member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_033409.3 | 2316 | UTR 3 | NP_212134.3 | |||
XM_005260655.3 | 2316 | UTR 3 | XP_005260712.1 | |||
XM_011529148.1 | 2316 | UTR 3 | XP_011527450.1 |