Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTAATTCACAAGCAAGTGCATTTG[A/G]TGGAAGAAGGCATCTTATGTGTTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609252 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LIPI PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LIPI - lipase I | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001302998.1 | 1426 | UTR 3 | NP_001289927.1 | |||
NM_001302999.1 | 1426 | UTR 3 | NP_001289928.1 | |||
NM_001303000.1 | 1426 | UTR 3 | NP_001289929.1 | |||
NM_001303001.1 | 1426 | Missense Mutation | ACC,ATC | T,I 370 | NP_001289930.1 | |
NM_198996.3 | 1426 | UTR 3 | NP_945347.2 | |||
XM_006723965.3 | 1426 | Intron | XP_006724028.1 |