Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAAGATTCACAGACAAGTGCAAAGG[A/G]GTGCTATTTTTAAAGGAGACCAAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606870 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
JAM2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
JAM2 - junctional adhesion molecule 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001270407.1 | Intron | NP_001257336.1 | ||||
NM_001270408.1 | Intron | NP_001257337.1 | ||||
NM_021219.3 | Intron | NP_067042.1 |