Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAACAGCACAAATTCTGGACTTTGA[A/G]TCTGGCTGCTGTCCTCACCTGAACC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605007 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADAMTS5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ADAMTS5 - ADAM metallopeptidase with thrombospondin type 1 motif 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_007038.4 | 7413 | UTR 3 | NP_008969.2 | |||
XM_011529448.2 | 7413 | UTR 3 | XP_011527750.1 |