Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGGTACCACAGGAATGGTGAGAAT[G/T]AACTTTTACAAGAGATGGTGAAAAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602490 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NRIP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NRIP1 - nuclear receptor interacting protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003489.3 | 5173 | UTR 3 | NP_003480.2 | |||
XM_005261063.3 | 5173 | UTR 3 | XP_005261120.1 | |||
XM_005261065.3 | 5173 | UTR 3 | XP_005261122.1 | |||
XM_011529747.1 | 5173 | UTR 3 | XP_011528049.1 | |||
XM_011529748.2 | 5173 | UTR 3 | XP_011528050.1 | |||
XM_011529749.2 | 5173 | UTR 3 | XP_011528051.1 | |||
XM_011529751.2 | 5173 | UTR 3 | XP_011528053.1 | |||
XM_011529752.1 | 5173 | UTR 3 | XP_011528054.1 | |||
XM_017028473.1 | 5173 | UTR 3 | XP_016883962.1 | |||
XM_017028474.1 | 5173 | UTR 3 | XP_016883963.1 | |||
XM_017028475.1 | 5173 | UTR 3 | XP_016883964.1 | |||
XM_017028476.1 | 5173 | UTR 3 | XP_016883965.1 |