Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCTCATCAACATGAGACACGGATT[C/T]GTAGGCAATTTCTGTACCTTTCATA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601245 MIM: 610078 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CHAF1B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CHAF1B - chromatin assembly factor 1 subunit B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005441.2 | 714 | Intron | NP_005432.1 | |||
XM_011529755.2 | 714 | Intron | XP_011528057.1 | |||
XM_017028477.1 | 714 | UTR 5 | XP_016883966.1 | |||
XM_017028478.1 | 714 | UTR 5 | XP_016883967.1 |
MORC3 - MORC family CW-type zinc finger 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |