Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTGCCACCAATGAGCGAGAGGGAC[G/T]AGGAGATGAAGCCCAGGTACAGGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605608 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CLDN14 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CLDN14 - claudin 14 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001146077.1 | 986 | Missense Mutation | NP_001139549.1 | |||
NM_001146078.2 | 986 | Missense Mutation | NP_001139550.1 | |||
NM_001146079.1 | 986 | Missense Mutation | NP_001139551.1 | |||
NM_012130.3 | 986 | Missense Mutation | NP_036262.1 | |||
NM_144492.2 | 986 | Missense Mutation | NP_652763.1 |