Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGGAGTTTCTACCCAAAGGCAGCC[G/T]TGCTCAGCCTCAACATCTTCCTTGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608617 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM3B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FAM3B - family with sequence similarity 3 member B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_058186.3 | Intron | NP_478066.3 | ||||
NM_206964.1 | Intron | NP_996847.1 | ||||
XM_011529648.2 | Intron | XP_011527950.2 | ||||
XM_011529649.2 | Intron | XP_011527951.1 |