Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCCCGAGAAGGGGGATGAGGTGAT[A/G]GTCCACAGGCCTCAGTGGACCGTCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 179020 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC105372824 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC105372824 - uncharacterized LOC105372824 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PDXK - pyridoxal (pyridoxine, vitamin B6) kinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003681.4 | Intron | NP_003672.1 | ||||
XM_005261195.2 | Intron | XP_005261252.2 | ||||
XM_005261196.3 | Intron | XP_005261253.1 | ||||
XM_005261198.2 | Intron | XP_005261255.1 | ||||
XM_005261199.2 | Intron | XP_005261256.1 | ||||
XM_011529758.2 | Intron | XP_011528060.1 | ||||
XM_011529760.2 | Intron | XP_011528062.1 | ||||
XM_011529761.1 | Intron | XP_011528063.1 | ||||
XM_011529762.2 | Intron | XP_011528064.1 | ||||
XM_017028483.1 | Intron | XP_016883972.1 | ||||
XM_017028484.1 | Intron | XP_016883973.1 |