Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCACCTGACGGAGGGCAGCTCACC[C/T]GCCTGCATCTATCCTCAATGCACTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610249 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LINC00334 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LINC00334 - long intergenic non-protein coding RNA 334 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
POFUT2 - protein O-fucosyltransferase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015227.4 | 1742 | UTR 3 | NP_056042.1 | |||
NM_133635.4 | 1742 | UTR 3 | NP_598368.2 | |||
XM_006723981.3 | 1742 | UTR 3 | XP_006724044.1 | |||
XM_011529504.2 | 1742 | UTR 3 | XP_011527806.1 | |||
XM_011529505.2 | 1742 | Intron | XP_011527807.1 | |||
XM_011529506.2 | 1742 | UTR 3 | XP_011527808.1 | |||
XM_011529507.2 | 1742 | UTR 3 | XP_011527809.1 | |||
XM_011529508.2 | 1742 | Intron | XP_011527810.1 | |||
XM_011529509.2 | 1742 | Intron | XP_011527811.1 | |||
XM_011529510.2 | 1742 | UTR 3 | XP_011527812.1 | |||
XM_011529511.2 | 1742 | Intron | XP_011527813.1 | |||
XM_011529512.2 | 1742 | Intron | XP_011527814.1 | |||
XM_011529513.2 | 1742 | Intron | XP_011527815.1 | |||
XM_017028306.1 | 1742 | UTR 3 | XP_016883795.1 | |||
XM_017028307.1 | 1742 | Intron | XP_016883796.1 | |||
XM_017028308.1 | 1742 | Intron | XP_016883797.1 | |||
XM_017028309.1 | 1742 | UTR 3 | XP_016883798.1 |