Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCCCCAGTGCATGTGGGATCCACG[A/G]CATGCTTCCTGCTTCGTCCTCTGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600594 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DGCR11 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DGCR11 - DiGeorge syndrome critical region gene 11 (non-protein coding) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
DGCR2 - DiGeorge syndrome critical region gene 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001173533.1 | 3546 | UTR 3 | NP_001167004.1 | |||
NM_001173534.1 | 3546 | UTR 3 | NP_001167005.1 | |||
NM_001184781.1 | 3546 | UTR 3 | NP_001171710.1 | |||
NM_005137.2 | 3546 | UTR 3 | NP_005128.1 |
DGCR5 - DiGeorge syndrome critical region gene 5 (non-protein coding) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |