Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGGGCCATCTGCTCCCAGGGCAGG[A/G]GACAGGCCACCAAGGACCTTTGGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607588 MIM: 608082 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PPIL2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PPIL2 - peptidylprolyl isomerase like 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317996.1 | 4119 | UTR 3 | NP_001304925.1 | |||
NM_014337.3 | 4119 | UTR 3 | NP_055152.1 | |||
NM_148175.2 | 4119 | UTR 3 | NP_680480.1 | |||
NM_148176.2 | 4119 | UTR 3 | NP_680481.1 | |||
XM_005261448.3 | 4119 | UTR 3 | XP_005261505.1 | |||
XM_011530041.2 | 4119 | UTR 3 | XP_011528343.1 | |||
XM_011530042.2 | 4119 | UTR 3 | XP_011528344.1 | |||
XM_011530043.2 | 4119 | UTR 3 | XP_011528345.1 | |||
XM_011530044.2 | 4119 | UTR 3 | XP_011528346.1 | |||
XM_011530046.2 | 4119 | UTR 3 | XP_011528348.1 | |||
XM_011530047.2 | 4119 | UTR 3 | XP_011528349.1 | |||
XM_011530049.2 | 4119 | UTR 3 | XP_011528351.1 | |||
XM_011530050.2 | 4119 | UTR 3 | XP_011528352.1 | |||
XM_011530051.2 | 4119 | UTR 3 | XP_011528353.1 | |||
XM_017028705.1 | 4119 | UTR 3 | XP_016884194.1 | |||
XM_017028706.1 | 4119 | UTR 3 | XP_016884195.1 |
YPEL1 - yippee like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_013313.4 | 4119 | UTR 3 | NP_037445.1 |