Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGCGCGACAGATCATGTGTCTTCT[C/T]TCTCACGAAGCTTACATTCTTGATG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600481 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCDC134 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CCDC134 - coiled-coil domain containing 134 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SREBF2 - sterol regulatory element binding transcription factor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004599.3 | Intron | NP_004590.2 | ||||
XM_006724310.2 | Intron | XP_006724373.1 | ||||
XM_011530347.1 | Intron | XP_011528649.1 | ||||
XM_017028921.1 | Intron | XP_016884410.1 | ||||
XM_017028922.1 | Intron | XP_016884411.1 |