Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCGCCCGCCACCGACTCACCTGCGC[C/T]GTTGTGGGGGGAGCCCCGGGGGAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600481 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SHISA8 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SHISA8 - shisa family member 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001207020.1 | 741 | Missense Mutation | AGC,GGC | S,G 220 | NP_001193949.1 | |
XM_006724254.3 | 741 | Missense Mutation | AGC,GGC | S,G 220 | XP_006724317.1 | |
XM_006724255.2 | 741 | Missense Mutation | AGC,GGC | S,G 220 | XP_006724318.1 | |
XM_006724256.3 | 741 | Missense Mutation | AGC,GGC | S,G 220 | XP_006724319.1 |
SREBF2 - sterol regulatory element binding transcription factor 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |