Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCCCTAATTCCCTCTCCTCCTGAC[G/T]CCTGTAGACATTCCCAATGATACCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608314 MIM: 610981 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SEPT3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SEPT3 - septin 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_019106.5 | 2583 | UTR 3 | NP_061979.3 | |||
NM_145733.2 | 2583 | Intron | NP_663786.2 | |||
XM_017028862.1 | 2583 | Intron | XP_016884351.1 |
WBP2NL - WBP2 N-terminal like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |