Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCTTAATTATCTCTTCCCGGATTT[C/T]TGTGAAAGCTTAGGAAATTTAACTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF280B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF280B - zinc finger protein 280B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_080764.3 | 4141 | UTR 3 | NP_542942.2 | |||
XM_011529893.2 | 4141 | UTR 3 | XP_011528195.1 | |||
XM_011529895.2 | 4141 | UTR 3 | XP_011528197.1 | |||
XM_011529896.2 | 4141 | UTR 3 | XP_011528198.1 | |||
XM_011529897.2 | 4141 | Intron | XP_011528199.1 |