Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCTGGCAGGCCAGCTCTGGTAGGT[A/G]ACTGTGTGGCAGAGCCCAGCCCCCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC107985555 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC107985555 - coiled-coil domain-containing protein 188-like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_017029168.1 | 677 | Missense Mutation | AAC,GAC | N,D 195 | XP_016884657.1 | |
XM_017029169.1 | 677 | Intron | XP_016884658.1 | |||
XM_017029170.1 | 677 | Intron | XP_016884659.1 |
ZDHHC8P1 - zinc finger DHHC-type containing 8 pseudogene 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |