Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCATTCATTCCTAGTCAACTCTAC[A/G]TCAATAACTGTTCCTGGAGGTGGAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610314 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PIWIL3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PIWIL3 - piwi like RNA-mediated gene silencing 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001008496.3 | 2782 | Silent Mutation | GAC,GAT | D,D 787 | NP_001008496.2 | |
NM_001255975.1 | 2782 | Silent Mutation | GAC,GAT | D,D 778 | NP_001242904.1 |