Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAAACTTGGGGCCAGGACATCCAA[A/T]ATGGGAATCCTCACTTACCAACGTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602254 MIM: 612747 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SRRD PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SRRD - SRR1 domain containing | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001013694.2 | 3538 | Intron | NP_001013716.2 | |||
XM_011530178.2 | 3538 | UTR 3 | XP_011528480.1 | |||
XM_017028799.1 | 3538 | Intron | XP_016884288.1 |
TFIP11 - tuftelin interacting protein 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001008697.1 | 3538 | Intron | NP_001008697.1 | |||
NM_012143.2 | 3538 | Intron | NP_036275.1 | |||
XM_011530079.1 | 3538 | Intron | XP_011528381.1 | |||
XM_017028735.1 | 3538 | Intron | XP_016884224.1 |