Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAACAGGATGATGGCTATAACCAG[G/T]TATCTGGTGGTCACTGGGGACTGTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608926 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
EMID1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EMID1 - EMI domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001267895.1 | 1411 | Intron | NP_001254824.1 | |||
NM_133455.3 | 1411 | Intron | NP_597712.2 | |||
XM_005261328.2 | 1411 | Intron | XP_005261385.1 | |||
XM_005261329.2 | 1411 | Intron | XP_005261386.1 | |||
XM_011529868.2 | 1411 | Intron | XP_011528170.1 | |||
XM_011529869.2 | 1411 | Intron | XP_011528171.1 | |||
XM_011529870.2 | 1411 | Intron | XP_011528172.1 | |||
XM_011529871.2 | 1411 | Intron | XP_011528173.1 | |||
XM_011529872.2 | 1411 | Intron | XP_011528174.1 | |||
XM_011529873.2 | 1411 | Intron | XP_011528175.1 | |||
XM_011529874.2 | 1411 | Intron | XP_011528176.1 | |||
XM_011529875.2 | 1411 | Intron | XP_011528177.1 | |||
XM_011529876.1 | 1411 | Intron | XP_011528178.1 | |||
XM_017028589.1 | 1411 | UTR 5 | XP_016884078.1 |