Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTAGACCTCCCAGACTCAGGAGATG[A/G]CACACAGCAGCTCAAGCACGAGCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603249 MIM: 612733 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NIPSNAP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NIPSNAP1 - nipsnap homolog 1 (C. elegans) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001202502.1 | Intron | NP_001189431.1 | ||||
NM_003634.3 | Intron | NP_003625.2 |
THOC5 - THO complex 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |