Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCATTCTGAGCCAGTGGGTGCCCA[A/G]CAATTCTATAAAGAAACCAGGATGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300883 | ||||||||||||||||||||
Literature Links: |
SNX12 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SNX12 - sorting nexin 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256185.1 | 534 | Missense Mutation | GCT,GTT | A,V 131 | NP_001243114.1 | |
NM_001256186.1 | 534 | Silent Mutation | TGC,TGT | C,C 85 | NP_001243115.1 | |
NM_001256187.1 | 534 | Silent Mutation | TGC,TGT | C,C 89 | NP_001243116.1 | |
NM_001256188.1 | 534 | Missense Mutation | GCT,GTT | A,V 127 | NP_001243117.1 | |
NM_013346.3 | 534 | Missense Mutation | GCT,GTT | A,V 131 | NP_037478.2 |