Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGAGTGATGTTACTGGCCCCCAAG[G/T]GTCTTGTTATATTACAGCACCATCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616979 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DTHD1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DTHD1 - death domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001136536.4 | 1091 | Missense Mutation | GGG,GTG | G,V 19 | NP_001130008.2 | |
NM_001170700.2 | 1091 | Missense Mutation | GGG,GTG | G,V 184 | NP_001164171.1 | |
XM_006714014.3 | 1091 | Missense Mutation | GGG,GTG | G,V 309 | XP_006714077.1 | |
XM_011513693.2 | 1091 | Missense Mutation | GGG,GTG | G,V 309 | XP_011511995.1 | |
XM_011513694.2 | 1091 | Missense Mutation | GGG,GTG | G,V 309 | XP_011511996.1 | |
XM_011513695.2 | 1091 | Missense Mutation | GGG,GTG | G,V 309 | XP_011511997.1 | |
XM_011513696.2 | 1091 | Missense Mutation | GGG,GTG | G,V 19 | XP_011511998.1 | |
XM_017008191.1 | 1091 | Missense Mutation | GGG,GTG | G,V 309 | XP_016863680.1 | |
XM_017008192.1 | 1091 | Missense Mutation | GGG,GTG | G,V 309 | XP_016863681.1 | |
XM_017008193.1 | 1091 | Missense Mutation | GGG,GTG | G,V 309 | XP_016863682.1 |