Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGACAAGGTCATGTGTGGATCTCCA[C/T]ACTCATCCTCTTCTCACTGATGCAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614829 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C4orf26 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C4orf26 - chromosome 4 open reading frame 26 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001206981.1 | Intron | NP_001193910.1 | ||||
NM_001257072.1 | Intron | NP_001244001.1 | ||||
NM_178497.3 | Intron | NP_848592.2 | ||||
XM_011531668.2 | Intron | XP_011529970.1 |