Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAGATCTAATTTTTATTGTTATACT[A/G]TTGTTGGTCGTGGTGATTATGAATA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610083 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TENM3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TENM3 - teneurin transmembrane protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080477.3 | Intron | NP_001073946.1 | ||||
XM_017008385.1 | Intron | XP_016863874.1 | ||||
XM_017008386.1 | Intron | XP_016863875.1 | ||||
XM_017008387.1 | Intron | XP_016863876.1 | ||||
XM_017008388.1 | Intron | XP_016863877.1 | ||||
XM_017008389.1 | Intron | XP_016863878.1 | ||||
XM_017008390.1 | Intron | XP_016863879.1 | ||||
XM_017008391.1 | Intron | XP_016863880.1 | ||||
XM_017008392.1 | Intron | XP_016863881.1 | ||||
XM_017008393.1 | Intron | XP_016863882.1 | ||||
XM_017008394.1 | Intron | XP_016863883.1 | ||||
XM_017008395.1 | Intron | XP_016863884.1 | ||||
XM_017008396.1 | Intron | XP_016863885.1 |