Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACAAGGCCTCCAGGTGATTCTGAT[A/G]TACACTAAAGCTTGAGGACCACTTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610586 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARHGAP24 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ARHGAP24 - Rho GTPase activating protein 24 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001025616.2 | Intron | NP_001020787.2 | ||||
NM_001042669.1 | Intron | NP_001036134.1 | ||||
NM_001287805.1 | Intron | NP_001274734.1 | ||||
NM_031305.2 | Intron | NP_112595.2 | ||||
XM_011532300.2 | Intron | XP_011530602.1 | ||||
XM_017008679.1 | Intron | XP_016864168.1 |