Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCTTCCAGATTCTCTTTGAGGAGC[A/G]ACTCTGTGTGTCAGTCTGACCCAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605912 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MEPE PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MEPE - matrix extracellular phosphoglycoprotein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001184694.2 | Intron | NP_001171623.1 | ||||
NM_001184695.1 | Intron | NP_001171624.1 | ||||
NM_001184696.1 | Intron | NP_001171625.1 | ||||
NM_001184697.1 | Intron | NP_001171626.1 | ||||
NM_001291183.1 | Intron | NP_001278112.1 | ||||
NM_020203.3 | Intron | NP_064588.1 | ||||
XM_006714278.2 | Intron | XP_006714341.1 |