Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTGAAGCTGAGGCTGAGTTGACCC[C/T]GAAAGCTTCTTGAGTAGATACAGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602368 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GRID2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GRID2 - glutamate ionotropic receptor delta type subunit 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286838.1 | Intron | NP_001273767.1 | ||||
NM_001510.3 | Intron | NP_001501.2 | ||||
XM_011531893.2 | Intron | XP_011530195.2 | ||||
XM_011531894.2 | Intron | XP_011530196.1 | ||||
XM_011531895.2 | Intron | XP_011530197.1 | ||||
XM_017008118.1 | Intron | XP_016863607.1 | ||||
XM_017008119.1 | Intron | XP_016863608.1 | ||||
XM_017008120.1 | Intron | XP_016863609.1 | ||||
XM_017008121.1 | Intron | XP_016863610.1 | ||||
XM_017008122.1 | Intron | XP_016863611.1 | ||||
XM_017008123.1 | Intron | XP_016863612.1 | ||||
XM_017008124.1 | Intron | XP_016863613.1 | ||||
XM_017008125.1 | Intron | XP_016863614.1 | ||||
XM_017008126.1 | Intron | XP_016863615.1 | ||||
XM_017008127.1 | Intron | XP_016863616.1 |