Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCATCGAAGGTTTGACTATGTCCTC[A/G]GTTGATACTCATAAATTTAAATATG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608350 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
EMCN PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EMCN - endomucin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001159694.1 | 3685 | UTR 3 | NP_001153166.1 | |||
NM_016242.3 | 3685 | UTR 3 | NP_057326.2 | |||
XM_011532024.2 | 3685 | Intron | XP_011530326.1 | |||
XM_017008290.1 | 3685 | Intron | XP_016863779.1 |
SNORA101A - small nucleolar RNA, H/ACA box 101A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |