Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTCTCCATGTGACCTTGCTGTGTC[A/C]AAAGCCACAGATCCTATTGCAGCCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611527 MIM: 611789 | ||||||||||||||||||||
Literature Links: |
SLC9B1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SLC9B1 - solute carrier family 9 member B1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC9B2 - solute carrier family 9 member B2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001300754.1 | 2047 | Missense Mutation | TTG,TTT | L,F 415 | NP_001287683.1 | |
NM_001300756.1 | 2047 | Nonsense Mutation | GGA,TGA | G,* 370 | NP_001287685.1 | |
NM_178833.5 | 2047 | Missense Mutation | TTG,TTT | L,F 472 | NP_849155.2 | |
XM_006714085.2 | 2047 | Missense Mutation | TTG,TTT | L,F 472 | XP_006714148.1 | |
XM_006714086.3 | 2047 | Missense Mutation | TTG,TTT | L,F 415 | XP_006714149.1 |