Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGAAACAGAATAAAAAAGCTTAGA[A/C]TCAAATAAGCATTGGGACACAGTCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
CNOT6L PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CNOT6L - CCR4-NOT transcription complex subunit 6 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286790.1 | 5880 | UTR 3 | NP_001273719.1 | |||
NM_144571.2 | 5880 | UTR 3 | NP_653172.2 | |||
XM_011531806.2 | 5880 | UTR 3 | XP_011530108.1 | |||
XM_011531807.2 | 5880 | UTR 3 | XP_011530109.1 | |||
XM_011531808.2 | 5880 | UTR 3 | XP_011530110.1 | |||
XM_011531809.1 | 5880 | UTR 3 | XP_011530111.1 | |||
XM_011531810.1 | 5880 | UTR 3 | XP_011530112.1 | |||
XM_017007956.1 | 5880 | UTR 3 | XP_016863445.1 | |||
XM_017007957.1 | 5880 | UTR 3 | XP_016863446.1 |