Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCAAGTTTAAAAAAGCAAAAAAAA[A/T]CGATCCTTTTGCACTGTTAAATAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 153245 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LEF1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LEF1 - lymphoid enhancer binding factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130713.2 | 2858 | UTR 3 | NP_001124185.1 | |||
NM_001130714.2 | 2858 | UTR 3 | NP_001124186.1 | |||
NM_001166119.1 | 2858 | UTR 3 | NP_001159591.1 | |||
NM_016269.4 | 2858 | UTR 3 | NP_057353.1 | |||
XM_005263046.3 | 2858 | UTR 3 | XP_005263103.1 | |||
XM_005263047.1 | 2858 | UTR 3 | XP_005263104.1 | |||
XM_005263048.1 | 2858 | UTR 3 | XP_005263105.1 | |||
XM_006714233.1 | 2858 | UTR 3 | XP_006714296.1 |