Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAGAAAATAGAATAAAAAATTTAA[T/C]ATTTCTCAAGTTGAACAAAACCCTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610851 MIM: 609028 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AP1AR PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AP1AR - adaptor related protein complex 1 associated regulatory protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TIFA - TRAF interacting protein with forkhead associated domain | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_052864.2 | 3056 | UTR 3 | NP_443096.1 |