Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTATTATACAAGTCTTTAAAAACT[A/G]CAATAAAGCATAGTGCCCCGATATT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601291 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
UGT8 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
UGT8 - UDP glycosyltransferase 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001128174.2 | Intron | NP_001121646.1 | ||||
NM_001322112.1 | Intron | NP_001309041.1 | ||||
NM_001322113.1 | Intron | NP_001309042.1 | ||||
NM_001322114.1 | Intron | NP_001309043.1 | ||||
NM_003360.4 | Intron | NP_003351.2 |