Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTTTATGGTACAATTGTACCTTAA[C/T]TGGCTTTATACCTGAGCACCAAAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607186 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SEC24D PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SEC24D - SEC24 homolog D, COPII coat complex component | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318066.1 | 2726 | UTR 3 | NP_001304995.1 | |||
NM_014822.3 | 2726 | UTR 3 | NP_055637.2 | |||
XM_005263379.2 | 2726 | UTR 3 | XP_005263436.1 | |||
XM_017008875.1 | 2726 | UTR 3 | XP_016864364.1 |