Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGTGAGTGAACAATCAACTCCAAA[A/G]TCCAGGAGAGCTGTCTAGATGGGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
C4orf3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C4orf3 - chromosome 4 open reading frame 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001001701.3 | Intron | NP_001001701.2 | ||||
NM_001170330.1 | Intron | NP_001163801.1 |
USP53 - ubiquitin specific peptidase 53 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |