Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTCTGTAGTAGAGCAGAAGGGGTC[C/T]AAAACCATGCACAATTTAACAGTCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603310 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC645513 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC645513 - septin 7 pseudogene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PDE5A - phosphodiesterase 5A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001083.3 | 6282 | UTR 3 | NP_001074.2 | |||
NM_033430.2 | 6282 | UTR 3 | NP_236914.2 | |||
NM_033437.3 | 6282 | UTR 3 | NP_246273.2 | |||
XM_017008791.1 | 6282 | Intron | XP_016864280.1 |