Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAAATATTTATTGATGGCTTACTAT[A/G]TGCTTTATGTCCTGTGTTAGATACT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
LINC01088 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LINC01088 - long intergenic non-protein coding RNA 1088 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NAA11 - N(alpha)-acetyltransferase 11, NatA catalytic subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032693.2 | Intron | NP_116082.1 | ||||
XM_017008777.1 | Intron | XP_016864266.1 | ||||
XM_017008778.1 | Intron | XP_016864267.1 | ||||
XM_017008779.1 | Intron | XP_016864268.1 |