Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTGCAGTGAACCCAGAAGAGGCTG[G/T]CGCCGACCTTAAGGGGCACTGTTTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
NAA11 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NAA11 - N(alpha)-acetyltransferase 11, NatA catalytic subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032693.2 | 129 | UTR 5 | NP_116082.1 | |||
XM_017008777.1 | 129 | UTR 5 | XP_016864266.1 | |||
XM_017008778.1 | 129 | UTR 5 | XP_016864267.1 | |||
XM_017008779.1 | 129 | UTR 5 | XP_016864268.1 |