Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTAAACGTGACTGTGCAAGACATT[A/C]ATGACAACCCCCCGGTTTTTGGCAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 612411 | ||||||||||||||||||||
Literature Links: |
FAT4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FAT4 - FAT atypical cadherin 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001291285.1 | 730 | Missense Mutation | AAT,CAT | N,H 244 | NP_001278214.1 | |
NM_001291303.1 | 730 | Missense Mutation | AAT,CAT | N,H 244 | NP_001278232.1 | |
NM_024582.4 | 730 | Missense Mutation | AAT,CAT | N,H 244 | NP_078858.4 | |
XM_011532236.2 | 730 | Missense Mutation | AAT,CAT | N,H 244 | XP_011530538.1 | |
XM_011532237.2 | 730 | Intron | XP_011530539.1 |