Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACCATATGAGCAGATATTATCACA[A/G]TCTTCCAGACAAGAAGCTGAAGCAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
HSPA4L PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HSPA4L - heat shock protein family A (Hsp70) member 4 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317381.1 | Intron | NP_001304310.1 | ||||
NM_001317382.1 | Intron | NP_001304311.1 | ||||
NM_001317383.1 | Intron | NP_001304312.1 | ||||
NM_014278.3 | Intron | NP_055093.2 | ||||
XM_011531745.2 | Intron | XP_011530047.1 |