Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATCAGCTACAGCCAATTTGTCAGT[C/G]ACGTGAGTAAACCACCTGGGACGTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
TMEM144 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TMEM144 - transmembrane protein 144 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018342.4 | 82 | Intron | NP_060812.2 | |||
XM_005263110.1 | 82 | Intron | XP_005263167.1 | |||
XM_005263112.3 | 82 | UTR 5 | XP_005263169.1 | |||
XM_006714254.1 | 82 | UTR 5 | XP_006714317.1 | |||
XM_017008366.1 | 82 | Intron | XP_016863855.1 | |||
XM_017008367.1 | 82 | Intron | XP_016863856.1 | |||
XM_017008368.1 | 82 | Intron | XP_016863857.1 |