Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAAGTCCCTGAATGTGCGAAGCGT[A/G]CAGTGCCTGCCCATACGCATCGCAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
9 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611501 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CAMTA1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CAMTA1 - calmodulin binding transcription activator 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001195563.1 | Intron | NP_001182492.1 | ||||
NM_001242701.1 | Intron | NP_001229630.1 | ||||
NM_015215.3 | Intron | NP_056030.1 | ||||
XM_011541083.2 | Intron | XP_011539385.1 | ||||
XM_011541084.2 | Intron | XP_011539386.1 | ||||
XM_011541085.2 | Intron | XP_011539387.1 | ||||
XM_011541086.2 | Intron | XP_011539388.1 | ||||
XM_011541087.2 | Intron | XP_011539389.1 | ||||
XM_011541088.2 | Intron | XP_011539390.1 | ||||
XM_011541089.2 | Intron | XP_011539391.1 | ||||
XM_011541090.2 | Intron | XP_011539392.1 | ||||
XM_011541091.2 | Intron | XP_011539393.1 | ||||
XM_011541092.2 | Intron | XP_011539394.1 | ||||
XM_017000773.1 | Intron | XP_016856262.1 | ||||
XM_017000774.1 | Intron | XP_016856263.1 | ||||
XM_017000775.1 | Intron | XP_016856264.1 | ||||
XM_017000776.1 | Intron | XP_016856265.1 | ||||
XM_017000777.1 | Intron | XP_016856266.1 | ||||
XM_017000778.1 | Intron | XP_016856267.1 | ||||
XM_017000779.1 | Intron | XP_016856268.1 | ||||
XM_017000780.1 | Intron | XP_016856269.1 | ||||
XM_017000781.1 | Intron | XP_016856270.1 |