Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTCCCACAACACAGTTTGAGGAAT[A/C]TGTTACTTTAAGCATCCTCTGTACT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
10 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605763 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC45A1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SLC45A1 - solute carrier family 45 member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080397.2 | Intron | NP_001073866.2 | ||||
XM_011541530.1 | Intron | XP_011539832.1 | ||||
XM_011541531.1 | Intron | XP_011539833.1 |